missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLITRK6 Antibody (sirtratumab) [Janelia Fluor™ 585], Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-28589JF585
This item is not returnable.
View return policy
Description
SLITRK6 Monoclonal antibody specifically detects SLITRK6 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| SLITRK6 | |
| Monoclonal | |
| Janelia Fluor 585 | |
| FLJ22774, MGC119595, MGC119596, MGC119597,4832410J21Rik, SLIT and NTRK-like family, member 6, SLIT and NTRK-like protein 6, slit and trk like gene 6 | |
| SLITRK6 | |
| 0.1 mL | |
| Neuroscience | |
| 84189 | |
| Store at 4°C in the dark. | |
| IgG2 |
| Flow Cytometry, ELISA, Functional Assay | |
| sirtratumab | |
| 50mM Sodium Borate | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction