missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLITRK6 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-00225-50ul
This item is not returnable.
View return policy
Description
SLITRK6 Polyclonal antibody specifically detects SLITRK6 in Human samples. It is validated for ELISA
Specifications
| SLITRK6 | |
| Polyclonal | |
| ELISA 1:5000-1:10000 | |
| FLJ22774, MGC119595, MGC119596, MGC119597,4832410J21Rik, SLIT and NTRK-like family, member 6, SLIT and NTRK-like protein 6, slit and trk like gene 6 | |
| Produced in rabbits immunized with purified, recombinant Human SLITRK6 (Uniprot#: Q9H5Y7; Met1-Ser608) | |
| 50 μg | |
| Neuroscience | |
| 84189 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction