missing translation for 'onlineSavingsMsg'
Learn More

SLITRK6 Antibody [DyLight 594], Novus Biologicals™

Product Code. 30731681 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30731681 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30731681 Supplier Novus Biologicals Supplier No. AF7317M

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

SLITRK6 Polyclonal antibody specifically detects SLITRK6 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLITRK6
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias FLJ22774, MGC119595, MGC119596, MGC119597,4832410J21Rik, SLIT and NTRK-like family, member 6, SLIT and NTRK-like protein 6, slit and trk like gene 6
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant human SLITRK6, Ile16-Ala604, Accession # Q9H5Y7
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 84189
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.