missing translation for 'onlineSavingsMsg'
Learn More

SLITRK5 Antibody, DyLight 680, Novus Biologicals™

Product Code. 18428985 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18428985 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18428985 Supplier Novus Biologicals Supplier No. NBP281914FR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SLITRK5 Polyclonal antibody specifically detects SLITRK5 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLITRK5
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 680
Formulation 50mM Sodium Borate
Gene Alias bA364G4.2, FLJ58374, KIAA0918, leucine-rich repeat-containing protein 11, LRRC11, SLIT and NTRK-like family, member 5, SLIT and NTRK-like protein 5
Host Species Rabbit
Immunogen SLITRK5 antibody was raised against a 14 amino acid synthetic peptide from near the center of human SLITRK5. The immunogen is located within amino acids 310 - 360 of SLITRK5. Amino Acid Squence: LKPPKGTRQPNKPR
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 26050
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.