missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLITRK4 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£141.00
Specifications
| Antigen | SLITRK4 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 0.5-1 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 2.5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 2.5 μg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
SLITRK4 Polyclonal specifically detects SLITRK4 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| SLITRK4 | |
| Western Blot 0.5-1 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 2.5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 2.5 μg/mL | |
| Polyclonal | |
| Rabbit | |
| Human | |
| Q8IW52 | |
| 139065 | |
| Antibody was raised against a 13 amino acid synthetic peptide from near the carboxy terminus of human Slitrk4. The immunogen is located within amino acids 760 - 810 of Slitrk4. Amino Acid Squence: PEKQPDKKSKKSL | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| PBS with 0.02% Sodium Azide | |
| DKFZp547M2010, SLIT and NTRK-like family, member 4, SLIT and NTRK-like protein 4, slit and trk like gene 4 | |
| SLITRK4 | |
| IgG | |
| Affinity Purified | |
| This antibody is predicted to have no cross-reactivity to other Slitrk proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title