missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SLITRK3 Polyclonal Antibody
GREENER_CHOICE

Product Code. 12848652
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12848652 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12848652 Supplier Invitrogen™ Supplier No. PA520474

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

A suggested positive control is SK-N-SH cell lysate. PA5-20474 can be used with blocking peptide PEP-0594.

SLIT and NTRK-like family 3 (Slitrk3) is a member a protein family consisting of six homologous transmembrane proteins (Slitrk1-6) that share two conserved leucine-rich repeat domains in the extracellular domain and have significant homology to Slit, a secreted axonal growth-controlling protein. These proteins are also homologous to trk neurotrophin receptors in their intracellular domains. Expression of Slitrk proteins is highly restricted to neural and brain tumor tissues, but varies within the protein family. Like every other Slitrk protein except Slitrk1, overexpression of Slitrk3 inhibited neurite outgrowth in cultured neurons, suggesting that these proteins are involved in the control of neurite outgrowth.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLITRK3
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene SLITRK3
Gene Accession No. O94933, Q810B9
Gene Alias KIAA0848; SLIT and NTRK like family member 3; SLIT and NTRK-like family, member 3; SLIT and NTRK-like protein 3; slit and trk like gene 3; SLITRK3
Gene Symbols SLITRK3
Host Species Rabbit
Immunogen A 14 amino acid peptide from near the carboxy terminus of human Slitrk3.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22865, 386750
Target Species Human, Mouse
Content And Storage Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.