missing translation for 'onlineSavingsMsg'
Learn More

SLITRK3 Antibody, mFluor Violet 450 SE, Novus Biologicals™

Product Code. 18428656 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18428656 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18428656 Supplier Novus Biologicals Supplier No. NBP281808MFV450

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SLITRK3 Polyclonal antibody specifically detects SLITRK3 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLITRK3
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias MGC138681, SLIT and NTRK-like family, member 3
Host Species Rabbit
Immunogen SLITRK3 antibody was raised against a 14 amino acid synthetic peptide from near the carboxy terminus of human SLITRK3. The immunogen is located within amino acids 780 - 830 of SLITRK3. Amino Acid Squence: FAETPKENHSNYRT
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 22865
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.