missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SLITRK2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 12818652
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12818652 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12818652 Supplier Invitrogen™ Supplier No. PA520471

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

A suggested positive control is rat brain tissue lysate. PA5-20471 can be used with blocking peptide PEP-0591.

This intronless gene is expressed in the hippocampus and maps close to a candidate region for several X-linked mental retardation (XLMR) syndromes. It is conserved in primates, cow, and horse, but not found in mouse and rat. The exact function of this gene is not known, but on the basis of its physical location and expression pattern, it is proposed to have an important function in the brain. [provided by RefSeq, Jan 2010].
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLITRK2
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene SLITRK2
Gene Accession No. Q810C0, Q9H156
Gene Alias A930040J07; CXorf2; KIAA1854; RGD1561361; SLIT and NTRK like family member 2; SLIT and NTRK-like family, member 2; SLIT and NTRK-like protein 2; slit and trk like gene 2; SLITL1; slit-like 1; Slitrk2; UNQ9197/PRO34756
Gene Symbols SLITRK2
Host Species Rabbit
Immunogen A 17 amino acid peptide from near the carboxy terminus of human Slitrk2.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 245450, 309349, 84631
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.