missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SLITRK2 Polyclonal Antibody
GREENER_CHOICE

Goat Polyclonal Antibody

Brand:  Invitrogen™ PA5148612

Product Code. 30534156

  • £399.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute at 0.2 mg/mL in sterile PBS.

This intronless gene is expressed in the hippocampus and maps close to a candidate region for several X-linked mental retardation (XLMR) syndromes. It is conserved in primates, cow, and horse, but not found in mouse and rat. The exact function of this gene is not known, but on the basis of its physical location and expression pattern, it is proposed to have an important function in the brain. [provided by RefSeq, Jan 2010].
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SLITRK2
Polyclonal
Unconjugated
SLITRK2
A930040J07; CXorf2; KIAA1854; RGD1561361; SLIT and NTRK like family member 2; SLIT and NTRK-like family, member 2; SLIT and NTRK-like protein 2; slit and trk like gene 2; SLITL1; slit-like 1; Slitrk2; UNQ9197/PRO34756
Goat
Antigen affinity chromatography
RUO
84631
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Western Blot
0.2 mg/mL
PBS with trehalose and no preservative
Q9H156
SLITRK2
Mouse myeloma cell line NS0-derived recombinant human SLITRK2, Thr19-Pro617.
100 μg
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.