missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLITRK2 Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-81841PCP
This item is not returnable.
View return policy
Description
SLITRK2 Polyclonal antibody specifically detects SLITRK2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| SLITRK2 | |
| Polyclonal | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin | |
| CXorf2, SLITL1, SLITRK2 SLIT and NTRK-like family, member 2 | |
| SLITRK2 antibody was raised against a 16 amino acid synthetic peptide from near the center of human SLITRK2. The immunogen is located within amino acids 650 - 700 of SLITRK2. Amino Acid Squence: LQYGSYNTETHDKTDG | |
| 0.1 mL | |
| Neuroscience | |
| 84631 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| PerCP | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction