missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLITRK1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£159.00 - £367.00
Specifications
| Antigen | SLITRK1 |
|---|---|
| Concentration | 0.65 mg/mL |
| Dilution | Western Blot 1:2500, Immunohistochemistry 1:250, Immunocytochemistry/Immunofluorescence 1:10-1:500 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18235002
|
Novus Biologicals
NBP1-49541SS |
0.025 mL |
£159.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18265854
|
Novus Biologicals
NBP1-49541 |
0.1 mL |
£367.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SLITRK1 Polyclonal specifically detects SLITRK1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
| SLITRK1 | |
| Western Blot 1:2500, Immunohistochemistry 1:250, Immunocytochemistry/Immunofluorescence 1:10-1:500 | |
| Polyclonal | |
| Rabbit | |
| Neuroscience | |
| PBS and 30% Glycerol with 0.05% Sodium Azide | |
| FLJ54428, KIAA1910KIAA0918, leucine rich repeat containing 12, Leucine-rich repeat-containing protein 12, LRRC12, RP11-395N17.1, SLIT and NTRK-like family, member 1, SLIT and NTRK-like protein 1, slit and trk like gene 1, TTM | |
| SLITRK1 | |
| IgG | |
| Affinity Purified | |
| Human and mouse. Immunogen sequence has 84% homology to rat. |
| 0.65 mg/mL | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human, Mouse | |
| Q96PX8, Q810C1 | |
| 114798 | |
| A genomic peptide made to an internal region of the human Slitrk1 protein (within residues 250-400). [Swiss-Prot Q96PX8] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title