missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLIT3 Monoclonal Antibody (782619)

Rat Monoclonal Antibody
Brand: Invitrogen™ MA561332
This item is not returnable.
View return policy
Description
Reconstitute with sterile PBS to a final concentration of 0.5 mg/mL.
Slit guidance ligand 3 (SLIT3) is a gene located on chromosome 5q35.1, encoding a member of the SLIT family of proteins that play key roles in axon guidance and neuronal migration during development. SLIT3 functions as a secreted protein, influencing the directional growth and pathfinding of axons by interacting with its receptor, Robo, on the surface of neurons. This interaction is critical for establishing neural network architecture and maintaining proper neural connectivity. Beyond its role in the nervous system, SLIT3 is involved in processes such as angiogenesis and cell migration, impacting tissue development and repair. Dysregulation of SLIT3 expression or function has been associated with developmental abnormalities and various diseases, including cancer, where it may influence tumor cell invasion and metastasis. Research into SLIT3 continues to explore its therapeutic potential in modulating axon guidance and its implications in pathological conditions affecting both the nervous and vascular systems.
Specifications
| SLIT3 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| Q9WVB4 | |
| SLIT3 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Slit3, Ser27-His901. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG2b |
| Immunohistochemistry (Frozen) | |
| 782619 | |
| Unconjugated | |
| SLIT3 | |
| b2b2362.1Clo; KIAA0814; Megf5; Multiple EGF-like domains protein 5; multiple epidermal growth factor-like domains 5; multiple epidermal growth factor-like domains protein 5; SLIL2; SLIL2FLJ10764; slit guidance ligand 3; slit homolog 3; slit homolog 3 (Drosophila); slit homolog 3 protein; SLIT1; slit2; SLIT3; Slit-3; UNQ691/PRO1336 | |
| Rat | |
| Protein A/G | |
| RUO | |
| 20564 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction