missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Slit3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-59140
This item is not returnable.
View return policy
Description
Slit3 Polyclonal specifically detects Slit3 in Human samples. It is validated for Western Blot.
Specifications
| Slit3 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| KIAA0814, MEGF5Multiple epidermal growth factor-like domains protein 5, Multiple EGF-like domains protein 5, SLIL2, SLIL2FLJ10764, slit homolog 3 (Drosophila), slit homolog 3 protein, SLIT1, slit2, Slit-3slit (Drosophila) homolog 3 | |
| Rabbit | |
| 168 kDa | |
| 100 μL | |
| Growth and Development, Neuronal Cell Markers | |
| 6586 | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| O75094 | |
| SLIT3 | |
| Synthetic peptides corresponding to SLIT3(slit homolog 3 (Drosophila)) The peptide sequence was selected from the N terminal of SLIT3 (NP_003053). Peptide sequence PRRLANKRISQIKSKKFRCSGSEDYRSRFSSECFMDLVCPEKCRCEGTIV. | |
| Affinity purified | |
| RUO | |
| Primary | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction