missing translation for 'onlineSavingsMsg'
Learn More

Slit3 Antibody (1D11), Novus Biologicals™

Product Code. 18326739 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18326739 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18326739 Supplier Novus Biologicals Supplier No. H00006586M07

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Slit3 Monoclonal antibody specifically detects Slit3 in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Slit3
Applications ELISA
Classification Monoclonal
Clone 1D11
Conjugate Unconjugated
Dilution ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_003053
Gene Alias KIAA0814, MEGF5Multiple epidermal growth factor-like domains protein 5, Multiple EGF-like domains protein 5, SLIL2, SLIL2FLJ10764, slit homolog 3 (Drosophila), slit homolog 3 protein, SLIT1, slit2, Slit-3slit (Drosophila) homolog 3
Host Species Mouse
Immunogen SLIT3 (NP_003053, 1371 a.a. ~ 1470 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. PCLGHRCHHGKCVATGTSYMCKCAEGYGGDLCDNKNDSANACSAFKCHHGQCHISDQGEPYCLCQPGFSGEHCQQENPCLGQVVREVIRRQKGYASCATA
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Growth and Development, Neuronal Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 6586
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.