Learn More
Invitrogen™ SLIT2 Recombinant Rabbit Monoclonal Antibody (24GB2210)
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557252
 View more versions of this product
Product Code. 30274884
  • £126.00 / 20µL
Description
SLIT2 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
SLIT2 are ligands for Robo receptors and play a vital role for axonal migration/navigation during neural development. A number of cleavage products through alternate splicing are reported in the literature for SLIT2 proteins. The C-terminal cleavage proteins are more diffusible than the larger N-terminal protein that is more tightly cell associated. SLIT2 protein is expressed in tissues such as fetal lung, kidney, and adult spinal cord.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SLIT2
Recombinant Monoclonal
Unconjugated
SLIT2
downregulated during adipocyte differentiation-1; Drad-1; E030015M03Rik; E130320P19Rik; mKIAA4141; neurogenic extracellular slit protein; SLIL3; Slit; slit guidance ligand 2; slit homolog 2 (Drosophila); slit homolog 2 protein; Slit homolog 2 protein C-product; Slit homolog 2 protein N-product; SLIT2; Slit-2; Slit2 protein
Rabbit
Affinity chromatography
RUO
20563, 360272, 9353
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
24GB2210
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
O94813, Q9R1B9, Q9WVC1
SLIT2
A synthesized peptide derived from human Slit2 (aa 1400-1500).
20 μL
Primary
Human, Mouse, Rat