missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Slit2 Antibody (02) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-26894
This item is not returnable.
View return policy
Description
Slit2 Monoclonal antibody specifically detects Slit2 in Human samples. It is validated for ELISA
Specifications
| Slit2 | |
| Monoclonal | |
| Unconjugated | |
| 0.2 μm filtered solution in PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 02 | |
| ELISA 1:1000-1:2000 | |
| FLJ14420, slit (Drosophila) homolog 2, slit homolog 2 (Drosophila), slit homolog 2 protein, Slit-2SLIL3 | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Slit2 (Uniprot#: O94813; aa 271-480). | |
| 100 μL | |
| Growth and Development, Neuronal Cell Markers, Neuroscience, Signal Transduction | |
| 9353 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction