missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLIT1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5121600
This item is not returnable.
View return policy
Description
SLIT1 Polyclonal Antibody for ICC/IF, IHC (P), Western Blot
SLIT1 is thought to act as molecular guidance cue in cellular migration, and function appears to be mediated by interaction with roundabout homolog receptors.
Specifications
| SLIT1 | |
| Polyclonal | |
| Unconjugated | |
| SLIT1 | |
| KIAA0813; Megf4; mKIAA0813; Multiple EGF-like domains protein 4; multiple epidermal growth factor-like domains 4; multiple epidermal growth factor-like domains protein 4; SLIL1; slit guidance ligand 1; slit homolog 1; slit homolog 1 (Drosophila); slit homolog 1 protein; SLIT1; Slit-1; SLIT3 | |
| Rabbit | |
| Protein A, Antigen affinity chromatography | |
| RUO | |
| 20562, 65047 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.86 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| O88279, Q80TR4 | |
| SLIT1 | |
| Recombinant protein Slit Homolog 1. The antigen corresponds to amino acid range 611-791 of the target protein. | |
| 20 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction