missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Slit1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB600-880
This item is not returnable.
View return policy
Description
Slit1 Polyclonal specifically detects Slit1 in Mouse, Rat samples. It is validated for Western Blot, ELISA.
Specifications
| Slit1 | |
| Polyclonal | |
| Unconjugated | |
| 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide | |
| KIAA0813, MEGF4slit (Drosophila) homolog 1, MGC164811, Multiple EGF-like domains protein 4, Multiple epidermal growth factor-like domains protein 4, SLIL1, slit homolog 1 (Drosophila), slit homolog 1 protein, Slit-1slit1, SLIT3 | |
| Rabbit | |
| 167.42 kDa | |
| 0.1 mg | |
| Growth and Development, Neuronal Cell Markers, Neuroscience, Signal Transduction | |
| 6585 | |
| Mouse, Rat | |
| IgG |
| Western Blot, ELISA | |
| 1.2 mg/ml | |
| Western Blot 1:500-1:2000, ELISA 1:10000-1:50000 | |
| Q80TR4, Q80TR4, Q80TR4, Q80TR4 | |
| SLIT1 | |
| This affinity purified Slit1 Antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to an internal region near aa 485-515 of mouse Slit1 protein. (Uniprot: Q80TR4) | |
| Affinity Purified | |
| RUO | |
| Primary | |
| SLIT1 | |
| Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction