Learn More
Invitrogen™ SLIRP Recombinant Rabbit Monoclonal Antibody (3H18L4)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ 703099
Description
This antibody is predicted to react with Monkey, Horse, Cat, Bovine. Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
SLIRP (SRA stem-loop interacting RNA binding protein) is expressed in normal and tumor tissues, but the majority of the endogenous SLIRP resides in the mitochondria. SLIRP may regulate mitochondrial functions and has the potential to shut down estrogen in breast cancer cells and testosterone in prostate cancer cells. SLIRP has also been shown to turn down genes involved in energy metabolism.
Specifications
| SLIRP | |
| Recombinant Monoclonal | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q9GZT3 | |
| SLIRP | |
| Protein corresponding to human SLIRP [aa19- aa103]. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot | |
| 3H18L4 | |
| Unconjugated | |
| SLIRP | |
| 1810035L17Rik; C14orf156; DC23; DC50; PD04872; SLIRP; SRA stem-loop interacting RNA binding protein; SRA stem-loop-interacting RNA-binding protein, mitochondrial | |
| Rabbit | |
| Protein A | |
| RUO | |
| 81892 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.