missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLIRP Polyclonal Antibody

SLIRP Polyclonal Antibody
Brand: Thermo Scientific PA5115923
This item is not returnable.
View return policy
Description
SLIRP Polyclonal Antibody for Western Blot, ICC/IF, IHC (PFA)
Specifications
| SLIRP | |
| Polyclonal | |
| Unconjugated | |
| SLIRP | |
| 1810035L17Rik; C14orf156; DC23; DC50; PD04872; SLIRP; SRA stem-loop interacting RNA binding protein; SRA stem-loop-interacting RNA-binding protein, mitochondrial | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 380773, 688717, 81892 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| Q9D8T7, Q9GZT3 | |
| SLIRP | |
| A synthesized peptide derived from human SLIRP(Accession Q9GZT3), corresponding to amino acid residues Q79-F109. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction