missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLIRP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA554175
This item is not returnable.
View return policy
Description
Immunogen sequence: LRRSINQPVA FVRRIPWTAA SSQLKEHFAQ FGHVRRCILP FDKETGFHRG LGWVQFSSEE GLRNALQQEN HIIDGVKVQV HTRRPKLPQT SDD Highest antigen sequence identity to the following orthologs: Mouse - 71%, Rat - 68%.
SLIRP (SRA stem-loop interacting RNA binding protein) is expressed in normal and tumor tissues, but the majority of the endogenous SLIRP resides in the mitochondria. SLIRP may regulate mitochondrial functions and has the potential to shut down estrogen in breast cancer cells and testosterone in prostate cancer cells. SLIRP has also been shown to turn down genes involved in energy metabolism.
Specifications
| SLIRP | |
| Polyclonal | |
| Unconjugated | |
| SLIRP | |
| 1810035L17Rik; C14orf156; DC23; DC50; PD04872; SLIRP; SRA stem-loop interacting RNA binding protein; SRA stem-loop-interacting RNA-binding protein, mitochondrial | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 81892 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9GZT3 | |
| SLIRP | |
| Recombinant protein corresponding to Human SLIRP. Recombinant protein control fragment (Product #RP-89130). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction