missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLIRP Antibody, DyLight 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB110-37258V
This item is not returnable.
View return policy
Description
SLIRP Polyclonal specifically detects SLIRP in Human, Mouse, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| SLIRP | |
| Polyclonal | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| Q9GZT3 | |
| SLIRP | |
| A synthetic peptide made to a C-terminal region of human SLIRP protein. [Entrez-Prot# AAX58600] | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse, Primate | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| DyLight 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| chromosome 14 open reading frame 156, DC50, PD04872, SLIRP, SRA stem-loop-interacting RNA-binding protein, mitochondrial | |
| Rabbit | |
| 12 kDa | |
| 0.1 mL | |
| Breast Cancer, Cancer, Chromatin Research, DNA replication Transcription Translation and Splicing, Signal Transduction, Stem Cell Markers | |
| 81892 | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction