missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLIRP Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB110-37258CL1
This item is not returnable.
View return policy
Description
SLIRP Polyclonal antibody specifically detects SLIRP in Human,Mouse,Primate samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Simple Western,Immunocytochemistry/Immunofluorescence
Specifications
| SLIRP | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Primate | |
| Purified |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Simple Western, Immunocytochemistry/Immunofluorescence | |
| CoraFluor 1 | |
| chromosome 14 open reading frame 156, DC50, PD04872, SLIRP, SRA stem-loop-interacting RNA-binding protein, mitochondrial | |
| A synthetic peptide made to a C-terminal region of human SLIRP protein. [Entrez-Prot# AAX58600] | |
| 0.1 mL | |
| Breast Cancer, Cancer, Chromatin Research, DNA replication Transcription Translation and Splicing, Signal Transduction, Stem Cell Markers | |
| 81892 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction