Learn More
Manutan Slip resistant tape, black, 18.3m x 102mm
Product Code. 12817398
Encompass_Preferred
Product Code. 12817398 Supplier Manutan Supplier No. A352779
Reduce the risk of slipping, promoting safety even in wet environments. Suitable for ladders, stair treads, ramps, walkways, entrance areas. Conforms to slip-resistant test EN 13552 Category R13.
Width : 102mm