Learn More
Thermo Scientific™ Sliding Drawer Inserts
Product Code. 17134205 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
13 cu. ft. Freezers
Type:
Microplate Insert
Unit Size:
Pack of 5
Product Code. For Use With (Equipment) Type unitSize
17134205 13 cu. ft. Freezers Microplate Insert Pack of 5
1 options
Product Code. 17134205 Supplier Thermo Scientific™ Supplier No. 1950666P5
Maximize storage space with these sliding drawer inserts, for use with racks used in Thermo Scientific™ 13, 17, 23 and 28 cu. ft. ultra-low temperature freezers.
Maximize storage space with Thermo Scientific™ Sliding Drawer Inserts, for use with racks used in Thermo Scientific™ 13, 17, 23 and 28 cu. ft. ultra-low temperature freezers.
  • Warranty varies by country, please contact us for details
TRUSTED_SUSTAINABILITY
Specifications
Type Microplate Insert
Capacity (English) 28 cu. ft.
Capacity (Metric) 792.8 L
For Use With (Equipment) 13 cu. ft. Freezers
Holds Microplate Rack