Learn More
Manutan Sliding door mesh cabinet, red, 1830mm x 915mm x 460mm
Description
A range of cabinets suitable for most work environments giving visibility of stock whilst maintaining security. Welded construction from 0. 9mm thick steel with reinforced mesh panel doors providing visibility and ventilation. Adjustable shelves, locks and keys supplied.
- Dark grey door
- Three shelves
- Dimensions (H x W x D) : 1830mm x 915mm x 460mm
- Capacity : 75kg
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.