missing translation for 'onlineSavingsMsg'
Learn More

Manutan Sliding door cabinet, red doors, 1020mm x 1830mm x 460mm

Product Code. 12821179
Encompass_Preferred
This item is not returnable. View return policy
Product Code. 12821179 Supplier Manutan Supplier No. A343194

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

The perfect space saving storage solution. Reinforced doors with adjustable shelves. Shelves adjustable in increments of 25mm with welded construction from 0. 9mm thick steel. Central lock supplied with 2 keys.

  • Capacity: 75kg
  • Colour of door: Red
  • Dimensions (H x W x D): 1020mm x 1830mm x 460mm
  • Two shelves
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.