missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manutan Sliding door cabinet, green doors, 1830mm x 1220mm x 460mm
Description
The perfect space saving storage solution. Reinforced doors with adjustable shelves. Shelves adjustable in increments of 25mm with welded construction from 0. 9mm thick steel. Central lock supplied with 2 keys.
- Capacity: 75kg
- Colour of door: Green
- Dimensions (H x W x D): 1830mm x 1220mm x 460mm
- Three shelves
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction