Learn More
CELLPATH SLIDERITE SUPA MEGA SLIDE - CARDBOARD MAILER
Product Code. 30226315
Change view
Click to view available options
Slide Capacity:
1
Unit Size:
Pack of 10
Product Code. Slide Capacity unitSize
30226315 1 Pack of 10
1 options
Product Code. 30226315 Supplier CELLPATH Supplier No. WEX050001A
  • ONLINE EXCLUSIVE PRICE
    £15.80
    £10.25 / Pack of 10
    Save £5.55
    35% Off
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
The robust cardboard mailers are constructed from a one-piece moulding that provides a high level of protection for Mega and Supa Mega slides.
  • Key Benefits:
  • Fits slides up to 76 x 52 mm
  • Economical
  • Re-usable
  • Tough and durable
Specifications
Type Slide Holder
Slide Capacity 1