Learn More
Zeiss™ Slider
Product Code. 12312043
Click to view available options
Type:
Slider Ph/PlasDIC, H, Ph/PlasDIC
Slider with Fixed Phase Stop Ph 1/0.4
Description:
Slider Ph/PlasDIC, H, Ph/PlasDIC
Slider w/ Fixed Phase Stop Ph 1/0.4
Unit Size:
Each
Product Code. 12312043
Brand: Zeiss™ 4267179020000
Only null left
Add to basket
Add to basket
Use these sliders for a variety of applications
Slider 10x46 mm Ph/PlasDIC, H, Ph/PlasDIC
  • 2 positions for Ph or PlasDIC diaphragms
  • brightfield position with neutral
  • density filter 0.06
Slider 10x46 mm
  • With fixed phase stop Ph 1/0.4
DIC slider EC EP
  • 5x/0.13, 10x/0.2, 20x/0.4 Epi+
DIC slider EC EP
  • 50x/0.7 Epi+
Specifications
Type Slider with Fixed Phase Stop Ph 1/0.4
Description Slider w/ Fixed Phase Stop Ph 1/0.4
Quantity 1 Ea.
Size 10 x 46 mm