missing translation for 'onlineSavingsMsg'
Learn More

Zeiss™ Slider

Product Code. 15360485
Change view
Click to view available options
Type:
Slider Ph/PlasDIC, H, Ph/PlasDIC
Slider with Fixed Phase Stop Ph 1/0.4
Description:
Slider Ph/PlasDIC, H, Ph/PlasDIC
Slider w/ Fixed Phase Stop Ph 1/0.4
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Description unitSize
15360485 Slider Ph/PlasDIC, H, Ph/PlasDIC Slider Ph/PlasDIC, H, Ph/PlasDIC Each
12312043 Slider with Fixed Phase Stop Ph 1/0.4 Slider w/ Fixed Phase Stop Ph 1/0.4 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15360485 Supplier Zeiss™ Supplier No. 4267179000000
  • ONLINE EXCLUSIVE PRICE
    £137.83
    £123.00 / Pack of 1
    Save £14.83
    10% Off
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Use these sliders for a variety of applications

Slider 10x46 mm Ph/PlasDIC, H, Ph/PlasDIC
  • 2 positions for Ph or PlasDIC diaphragms
  • brightfield position with neutral
  • density filter 0.06
Slider 10x46 mm
  • With fixed phase stop Ph 1/0.4
DIC slider EC EP
  • 5x/0.13, 10x/0.2, 20x/0.4 Epi+
DIC slider EC EP
  • 50x/0.7 Epi+

Specifications

Type Slider Ph/PlasDIC, H, Ph/PlasDIC
Description Slider Ph/PlasDIC, H, Ph/PlasDIC
For Use With (Application) Ph or PlasDIC Diaphragms
Quantity 1 Ea.
Size 10 x 46 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.