Learn More
Epredia™ SlideMate™ Accessories
Product Code. 12943815 Shop All Epredia Products
Click to view available options
Product Type:
Digital Imager
Keyboard
Keyboard Interface
LabWriter Software
Mini PC Package, clamp arm
Mini PC Package, stand
Mini PC, imager, clmp arm
Mini PC, imager, stand
Thermal Transfer Ribbon
Unit Size:
Each
Product Code. 12943815
Brand: Epredia™ B81300031
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Enhance and optimize the Epredia™ SlideMate™ Slide Printer with Epredia™ SlideMate Slide Printer Accessories.
Accessories include:
  • Collection system Collection trays (box of 4)
  • Hopper
  • Hot foil tape, box of 5 (roll of tape prints approx. 20,000 cassettes)
TRUSTED_SUSTAINABILITY
Specifications
Product Type Mini PC Package, stand
For Use With (Equipment) SlideMate™ Slide Printer
Includes Desktop Stand