Learn More
BRAND™ Slide Storage Box
Shop All Brand Products
Click to view available options
:
Material: Polypropylene
Description
- Sturdy design
- Screw lid with dealing wire lug
- Ideal for storing or shipping 5 thick or 10 thin slides
- Slides protrude 10 mm above the box to facilitate removal
Specifications
Specifications
| For Use With (Application) | Slide storage and transport |
| For Use With (Equipment) | Slide |
| Material | Polypropylene |