missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Labnet™ Slide Spinner
Description
- Ideal for removing wash buffer from microarrays prior to scanning
- Removable cassettes conveniently catch all liquid
- Quickly dries 2 slides
- Designed for drying liquid from glass slides, typically microarray slides
Specifications
Specifications
| Capacity | 2 Standard Slides |
| Max. Speed | 4,800 rpm |
| Type | Slide Spinner |
| Dimensions (HWD) | 11.7 x 15 x 15 cm (4.6 x 5.9 x 5.9 in.) |
| Frequency | 50 Hz |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?