Learn More
Epredia™ Slide Baskets and Hangers for Shandon™ ClearVue™ Coverslippers, Sakura DRS 2000-compatible baskets (white sliders)
Product Code. 12658196 Shop All Epredia Products
Product Code. 12658196
Brand: Epredia™ A79210067
Use these slide baskets and hangers with the Epredia™ Shandon™ ClearVue™ Coverslipper.
  • Allow equipment compatibility with a variety of different stainers
  • Can be loaded in mix-or-match fashion
  • Sold in sets of five
TRUSTED_SUSTAINABILITY
Specifications
Product Type With White Sliders
For Use With (Equipment) Shandon ClearVue Coverslipper
Description Sakura DRS 2000-Compatible Baskets