Learn More
Epredia™ Slide Baskets and Hangers for Shandon™ ClearVue™ Coverslippers, Leica baskets (White sliders)
Use these slide baskets and hangers with the Epredia™ Shandon™ ClearVue™ Coverslipper.
Brand: Epredia™ A79210073
Code : 50
Additional Details : Weight : 1.00000kg
Description
- Allow equipment compatibility with a variety of different stainers
- Can be loaded in mix-or-match fashion
- Sold in sets of five
Specifications
With White Sliders | |
Shandon ClearVue Coverslipper | |
Leica Baskets |