Learn More
Epredia™ Slide Baskets and Hangers for Epredia™ ClearVue™ Coverslipper
Product Code. 12628196 Shop All Epredia Products
Click to view available options
Product Type:
Gemini Basket, Black Slider
Gemini Basket, White Slider
Leica Basket, Black Slider
Unit Size:
Pack of 5
Product Code. 12628196
Brand: Epredia™ A79210064
Use these slide baskets and hangers with the Epredia™ ClearVue™ Coverslipper to make it compatible with any automated slide stainer.
  • Allow equipment compatibility with a variety of different stainers
  • Can be loaded in mix-or-match fashion
  • Sold in sets of five
TRUSTED_SUSTAINABILITY
Specifications
Product Type Gemini Basket, Black Slider
For Use With (Equipment) Epredia ClearVue Coverslipper
Quantity 5 Pk.