Learn More
Thermo Scientific™ Slide-A-Lyzer™ G2 Dialysis Cassettes, 10K MWCO
Shop All Thermo Scientific ProductsDescription
Slide-A-Lyzer G2 Dialysis Cassettes (10K MWCO) provide maximum convenience, flexibility, and performance for sample dialysis. They allow samples to be loaded with a syringe or pipette through the unique sample port. The sample is easily recovered through either the pipette or syringe sample ports. The Slide-A-Lyzer Cassette design allows air to be removed from inside the cassette to maximize the surface area-to-volume ratio, providing better dialysis performance than dialysis tubing and tube-based systems.
The 10K molecular-weight cutoff (MWCO) cassettes use a low-binding regenerated cellulose that is rated to completely retain proteins and other macromolecules larger than 10,000 Daltons while allowing removal of buffer salts and small contaminants. Membranes and cassettes are manufactured using clean-room conditions to ensure that are contaminant-free.
Features of Slide-A-Lyzer G2 Dialysis Cassettes (10K MWCO):
• Easy-loading—pipette-accessible for easy sample loading and retrieval
• Self-floating—integrated air chambers eliminate the need for float buoys
• Sturdy construction—ensures the highest possible sample integrity and protection
• Superior design—thoroughly researched and tested to provide fast and consistent dialysis with maximum sample recovery
• Versatile—ideal for removing low molecular-weight contaminants, performing buffer exchange and desalting
• Multiple sizes—five cassette capacities to optimally match 0.5 to 70 mL sample volumes
• Gamma-irradiated option—gamma-irradiated cassettes are available for applications requiring conditions that minimize the risk of sample contamination
Gamma-irradiated (sterile) dialysis cassettes
Gamma-irradiated Slide-A-Lyzer G2 Dialysis Cassettes are ideal for researchers culturing cells and microorganisms; purifying viruses, DNA, and RNA; or performing sample preparation for other applications requiring conditions that minimize the risk of sample contamination. The gamma-irradiated cassettes are treated with 20–36 kGy of gamma radiation, a dosage that has been determined to sterilize the units based on both trypticase soy broth medium for identifying total growth and fluid thioglycollate medium for identifying anaerobic growth. However, a confirmation of sterility is not provided for individual lots, as no further sterility testing is conducted post irradiation.
Membrane specifications:
• Composition: regenerated cellulose synthesized by the viscose method
• Thickness: 0.9 to 1.2 mil (22.5 to 30 μm)
• Hydration time required before use: 2 minutes
• Glycerol content: 21%
• Sulfur content: 0.1–0.15%
• Heavy metals content: trace
Volume ranges:
• 0.5 mL = 0.25 to 0.75 mL
• 3 mL = 1 to 3 mL
• 15 mL = 5 to 15 mL
• 30 mL = 10 to 30 mL
• 70 mL = 25 to 70 mL
Order Info
Important Note: SKU #87729 will be discontinued and replaced by 66383, 66384, or 88404 by June 1, 2023. Consider switching to 66383, 66384, or 88404.
Specifications
Specifications
| Content And Storage | Store at room temperature |
| Purification Target | Buffer Exchange, Protein |
| Membrane | Cellulose Membrane, Dialysis |
| Format | Cassette |
| MWCO | 10 kDa |
| Product Line | Slide-A-Lyzer |
| Quantity | 10 Cassettes |
| Sample Volume (Metric) | 0.5 mL |
| Product Type | G2 Dialysis Cassettes |
Frequently Asked Questions (FAQs)
It is possible to add reagents to the Slide-A-Lyzer Dialysis Cassette at different times using different portholes, allow a reaction to occur, and then dialyze in the same Slide-A-Lyzer Dialysis Cassette, reducing sample loss through reduced sample handling.
The Slide-A-Lyzer Dialysis Cassette is the same regenerated cellulose as other dialysis tubing so you can expect about the same amount of protein loss as with tubing. As a guide, a 1 mg/mL solution will have a recovery rate of greater than 95%; at 100 mg/mL the recovery drops to 75-80%; and in solutions as dilute 10 µg/mL users may observe only a 50% (or less) recovery.
Dialyze for the amount of time sufficient to remove low molecular weight compounds for the specific downstream application. A typical dialysis procedure is as follows:
1) Dialyze for 2 hrs at room temperature or 4 degrees C.
2) Change the dialysis buffer and dialyze for another 2 hrs.
3) Change the dialysis buffer and dialyze overnight at 4 degrees C.
Use the dialysis buffer at 200-500 times the volume of the sample. For example, if the starting sample volume is 1 mL, then use a dialysis buffer amount that is equal to 200 mL-500 mL for each dialysis buffer change.
The buoy plays two roles: it suspends the Slide-A-Lyzer Dialysis Cassette above the stirring bar during dialysis; it provides a stand to set the Slide-A-Lyzer Dialysis Cassette in on the bench-top
No, any 18 gauge needle at least one inch long, and syringes of sufficient volume will work.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.