missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Slide-A-Lyzer™ Dialysis Cassettes, 7K MWCO

Código de producto. 10761195 Tienda Thermo Scientific Productos
Cambiar vista
Click to view available options
Sample Volume (Metric):
0.5 mL
3 mL
12 mL
Quantity:
1 Kit (10 Cassettes)
1 Kit (8 Cassettes)
10 Cassettes
8 Cassettes
Tamaño de la unidad:
Pack of 10
Pack of 8
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Sample Volume (Metric) Quantity Tamaño de la unidad
10761195 12 mL 8 Cassettes Pack of 8
10250054 0.5 mL 10 Cassettes Pack of 10
10240294 3 mL 10 Cassettes Pack of 10
10415525 3 mL 1 Kit (10 Cassettes) Pack of 10
10608854 12 mL 1 Kit (8 Cassettes) Pack of 8
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 10761195 Proveedor Thermo Scientific™ N.º de proveedor 66710

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Ver productos alternativos

missing translation for 'validMainframeMsg'

Este artículo no se puede devolver. Vea la política de devoluciones

Remove salts and contaminants and maximize sample recovery using this semi-permeable membrane with a 7,000-Dalton protein MW cut-off (7K MWCO).

Slide-A-Lyzer Dialysis Cassettes (7K MWCO) facilitate the simple and effective removal of salts and small contaminants from proteins or other macromolecules greater than 7,000 Daltons. They are available in four sizes to accommodate sample volumes between 0.5 and 12 mL. These devices are easier to use than traditional dialysis tubing, providing faster buffer exchange, and more complete sample recovery.

Slide-A-Lyzer cassettes can be used for a wide range of applications including low-molecular weight contaminant removal, buffer exchange, desalting, equilibrium dialysis, and sample concentration. Liquid samples are easily added and removed by penetrating the self-sealing gasket with a hypodermic needle attached to a syringe. No knots, clips, or caps are needed to seal the units and prevent leaking and sample-loss during the dialysis procedure.

Slide-A-Lyzer Dialysis Cassettes are constructed from two sheets of low-binding, regenerated-cellulose dialysis membrane that are hermetically sealed on either side of a silicone-like gasket inside an inert plastic frame. The membrane and cassette materials are compatible with most common laboratory chemicals and buffers.

Features of Slide-A-Lyzer Dialysis Cassettes:
• Easy to use—no knots or clamps are needed; just inject sample into cassette and begin dialysis
• Fast dialysis—flat cassette chamber with two membranes provides high surface-area to volume ratio that maximizes diffusion rate compared to cylindrical dialysis tubing
• High recovery—rectangular cassette design maximizes recovery of entire sample volume via any one of the four corner injection ports
• Four cassette sizes—select the cassette that best suits your sample volume
• Color-coded—green cassette frame specifies cassettes with the 7,000 MW pore size membrane

Membrane specifications:
• Composition: regenerated cellulose synthesized by the viscose method
• Thickness: 0.9 to 1.2 mil (22.5 to 30 μm)
• Hydration time required before use: 2 minutes
• Glycerol content: 13%
• Sulfur content: 0.1 to 0.15%
• Heavy metals content: trace

Volume ranges:
• 0.5 mL = 0.1 to 0.5 mL
• 3 mL = 0.5 to 3 mL
• 12 mL = 3 to 12 mL

TRUSTED_SUSTAINABILITY

Especificaciones

Content And Storage Store at room temperature
Purification Target Buffer Exchange, Protein
Membrane Cellulose Membrane, Dialysis
Format Cassette
MWCO 7.0 kDa
Product Line Slide-A-Lyzer
Quantity 8 Cassettes
Sample Volume (Metric) 12 mL

Slide-A-Lyzer Dialysis Cassette Technology is protected by U.S. Patent 5,503,741.

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.