missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Slide-A-Lyzer™ Dialysis Cassettes, 20K MWCO

Product Code. 10260404 Shop All Thermo Scientific Products
Change view
Click to view available options
Sample Volume (Metric):
0.5 mL
3 mL
12 mL
30 mL
Unit Size:
Pack of 10
Pack of 6
Pack of 8
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sample Volume (Metric) unitSize
10260404 30 mL Pack of 6
10668884 0.5 mL Pack of 10
10771775 3 mL Pack of 10
10200364 12 mL Pack of 8
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10260404 Supplier Thermo Scientific™ Supplier No. 66030

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Use this 20K MWCO semi-permeable membrane to remove salts and contaminants with maximal sample recovery.

Slide-A-Lyzer Dialysis Cassettes (20K MWCO) facilitate the simple and effective removal of buffer salts and small contaminants from proteins and other macromolecules larger than 20,000 Daltons. They are available in four sizes to accommodate sample volumes between 0.5 and 30 mL. These devices are easier to use than traditional dialysis tubing, providing faster buffer exchange, and more complete sample recovery.

Slide-A-Lyzer Dialysis Cassettes (20K MWCO) facilitate the simple and effective removal of buffer salts and small contaminants from proteins and other macromolecules larger than 20,000 Daltons. They are available in four sizes to accommodate sample volumes between 0.5 and 30 mL. These devices are easier to use than traditional dialysis tubing, providing faster buffer exchange and more complete sample recovery. Slide-A-Lyzer cassettes can be used for a wide range of applications including low-molecular weight contaminant removal, buffer exchange, desalting, equilibrium dialysis, and sample concentration. Liquid samples are easily added and removed by penetrating the self-sealing gasket with a hypodermic needle attached to a syringe. No knots, clips, or caps are needed to seal the units and prevent leaking and sample-loss during the dialysis procedure.

Slide-A-Lyzer Dialysis Cassettes are constructed from two sheets of low-binding, regenerated-cellulose dialysis membrane that are hermetically sealed on either side of a silicone-like gasket inside an inert plastic frame. The membrane and cassette materials are compatible with most common laboratory chemicals and buffers.

Features of Slide-A-Lyzer Dialysis Cassettes:

  • Easy to use—no knots or clamps are needed; just inject sample into cassette and begin dialysis
  • Fast dialysis—flat cassette chamber with two membranes provides high surface-area to volume ratio that maximizes diffusion rate compared to cylindrical dialysis tubing
  • High recovery—rectangular cassette design maximizes recovery of entire sample volume via any one of the four corner injection ports
  • Four cassette sizes—select the cassette that best suits your sample volume
  • Color-coded—purple cassette frame specifies cassettes with the 20,000 MW pore size membrane

Membrane specifications:

  • Composition: regenerated cellulose synthesized by the Viscose method
  • Thickness: 0.9 to 1.2 mil (22.5 to 30 μm)
  • Hydration time required before use: 2 minutes
  • Glycerol content: none
  • Sulfur content: 0.04%
  • Heavy metals content: trace

Volume ranges:

  • 0.5 mL = 0.1 to 0.5 mL
  • 3 mL = 0.5 to 3 mL
  • 12 mL = 3 to 12 mL
  • 30 mL = 12 to 30 mL
TRUSTED_SUSTAINABILITY

Specifications

Description Slide-A-Lyzer Dialysis Cassettes, 20K MWCO, 30mL
Content And Storage Store at room temperature
Purification Target Buffer Exchange, Protein
Membrane Cellulose Membrane, Dialysis
Format Cassette
MWCO 20 kDa
Product Line Slide-A-Lyzer
Quantity 6 Cassettes
Sample Volume (Metric) 30 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.