missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Slide-A-Lyzer™ G2 Dialysis Cassettes, 20K MWCO

Product Code. 10226814 Shop All Thermo Scientific Products
Change view
Click to view available options
Sample Volume (Metric):
0.5 mL
3 mL
15 mL
30 mL
70 mL
Unit Size:
Pack of 10
Pack of 6
Pack of 8
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sample Volume (Metric) unitSize
10226814 30 mL Pack of 6
10627575 0.5 mL Pack of 10
10216814 3 mL Pack of 10
10449844 15 mL Pack of 8
10402145 70 mL Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 10226814 Supplier Thermo Scientific™ Supplier No. 87737

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Important Note: These G2 Dialysis Cassettes will be discontinued on June 1, 2023 and replaced by G3 Dialysis Cassettes, a new generation of cassettes. Consider switching to G3 Dialysis Cassettes. Remove salts and contaminants while maximizing sample recovery with these second-generation, self-floating protein dialysis cassettes.

Slide-A-Lyzer G2 Dialysis Cassettes (20K MWCO) provide maximum convenience, flexibility, and performance for sample dialysis.

Slide-A-Lyzer G2 Dialysis Cassettes allow samples to be directly injected with a syringe or pipette through the unique sample port. The cassette design allows air to be removed from inside the cassette to maximize the surface area-to-volume ratio, providing better dialysis performance than dialysis tubing and tube-based systems. The sample is easily recovered through either the pipette or syringe sample ports.

The 20K molecular-weight cut off (MWCO) cassettes use a cellulose membrane that is rated to completely retain proteins and other macromolecules larger than 20,000 Daltons while allowing removal of buffer salts and small contaminants. The cassette membrane is composed of low-binding regenerated cellulose. Membranes and cassettes are manufactured using clean-room conditions to ensure they are contaminant-free.

Features of Slide-A-Lyzer G2 Dialysis Cassettes (20K MWCO):
Easy-loading—pipette-accessible for easy sample loading and retrieval
Self-floating—integrated air chambers eliminate the need for float buoys
Sturdy construction—ensures the highest possible sample integrity and protection
Superior design—thoroughly researched and tested to provide fast and consistent dialysis with maximum sample recovery
Multiple sizes—cassette capacities to optimally match 0.5 to 70 mL sample volumes
Versatile—ideal for removing low-molecular weight contaminants, performing buffer exchange and desalting

Volume ranges:
• 0.5 mL = 0.25 to 0.75 mL
• 3 mL = 1 to 3 mL
• 15 mL = 5 to 15 mL
• 30 mL = 10 to 30 mL
• 70 mL = 25 to 70 mL

Order Info

Important Note: SKU #87737 will be discontinued on June 1, 2023 and replaced by A52978, a new generation of cassettes. Consider switching to A52978.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at room temperature
Purification Target Buffer Exchange, Protein
Membrane Cellulose Membrane, Dialysis
Format Cassette
MWCO 20 kDa
Product Line Slide-A-Lyzer
Quantity 6 Cassettes
Sample Volume (Metric) 30 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.