missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Slide-A-Lyzer™ Dialysis Cassettes, 10K MWCO

Product Code. 10167913 Shop All Thermo Scientific Products
Change view
Click to view available options
Sample Volume (Metric):
0.5 mL
3 mL
12 mL
30 mL
Quantity:
10 Cassettes
5 x 10 Cassettes
5 x 8 Cassettes
6 Cassettes
8 Cassettes
Product Type:
Dialysis Cassette Kit
Dialysis Cassettes
Dialysis Cassettes (gamma-irradiated)
Unit Size:
Pack of 10
Pack of 40
Pack of 50
Pack of 6
Pack of 8
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sample Volume (Metric) Quantity Product Type unitSize
10167913 30 mL 6 Cassettes Dialysis Cassettes Pack of 6
10194183 0.5 mL 10 Cassettes Dialysis Cassettes Pack of 10
10731765 0.5 mL 5 x 10 Cassettes Dialysis Cassettes Pack of 50
10065983 0.5 mL 10 Cassettes Dialysis Cassette Kit Pack of 10
10591004 3 mL 10 Cassettes Dialysis Cassettes Pack of 10
10678464 3 mL 5 x 10 Cassettes Dialysis Cassettes Pack of 50
11889943 3 mL 10 Cassettes Dialysis Cassette Kit Pack of 10
10539853 12 mL 8 Cassettes Dialysis Cassettes Pack of 8
10311014 12 mL 5 x 8 Cassettes Dialysis Cassettes Pack of 40
10591474 12 mL 8 Cassettes Dialysis Cassette Kit Pack of 8
10311364 0.5 mL 10 Cassettes Dialysis Cassettes (gamma-irradiated) Pack of 10
10371404 3 mL 10 Cassettes Dialysis Cassettes (gamma-irradiated) Pack of 10
10301554 12 mL 8 Cassettes Dialysis Cassettes (gamma-irradiated) Pack of 8
10076033 30 mL 6 Cassettes Dialysis Cassettes (gamma-irradiated) Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
This item is not returnable. View return policy
Product Code. 10167913 Supplier Thermo Scientific™ Supplier No. 66830

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Remove salts and small contaminants to maximize sample purity and recovery with Slide-A-Lyzer 10K MWCO dialysis cassettes.

Slide-A-Lyzer Dialysis Cassettes (10K MWCO) facilitate the simple and effective removal of salts and small contaminants from proteins and other macromolecules greater than 10,000 Daltons. Slide-A-Lyzer Dialysis Cassettes are available in four sizes to accommodate sample volumes between 0.5 and 30 mL. These devices are easier to use than traditional dialysis tubing, providing faster buffer exchange and more complete sample recovery.

Slide-A-Lyzer cassettes can be used for a wide range of applications including low-molecular weight contaminant removal, buffer exchange, desalting, equilibrium dialysis, and sample concentration. Liquid samples are easily added and removed by penetrating the self-sealing gasket with a hypodermic needle attached to a syringe. No knots, clips, or caps are needed to seal the units and prevent leaking and sample-loss during the dialysis procedure.

Slide-A-Lyzer Dialysis Cassettes are constructed from two sheets of low-binding, regenerated-cellulose dialysis membrane that are hermetically sealed on either side of a silicone-like gasket inside an inert plastic frame. The membrane and cassette materials are compatible with most common laboratory chemicals and buffers.

Features of Slide-A-Lyzer Dialysis Cassettes:

  • Color-coded—orange frame specifies cassettes with the 10,000 MW pore size membrane
  • Four cassette sizes—select the cassette that best suits your sample volume
  • Fast dialysis—flat cassette chamber with two membranes provides high surface area-to-volume ratio that maximizes diffusion rate compared to cylindrical dialysis tubing
  • High recovery—rectangular cassette design maximizes recovery of entire sample volume via any one of the four corner injection ports
  • Gamma-irradiated option—gamma-irradiated cassettes are available for applications requiring conditions that minimize the risk of sample contamination

Gamma-irradiated dialysis cassettes

Gamma-irradiated Slide-A-Lyzer Dialysis Cassettes are ideal for researchers culturing cells and microorganisms; purifying viruses, DNA, and RNA; or performing sample preparation for other applications requiring conditions that minimize the risk of sample contamination. The gamma-irradiated cassettes are treated with 20–36 kGy of gamma radiation, a dosage that has been determined to sterilize the units based on both trypticase soy broth medium for identifying total growth and fluid thioglycollate medium for identifying anaerobic growth. However, a confirmation of sterility is not provided for individual lots, as no further sterility testing is conducted post-irradiation.

Membrane specifications:

  • Composition: regenerated cellulose synthesized by the viscose method
  • Thickness: 0.9 to 1.2 mil (22.5 to 30 μm)
  • Hydration time required before use: 2 minutes
  • Glycerol content: 21%
  • Sulfur content: 0.1–0.15%
  • Heavy metals content: trace

Volume ranges:

  • 0.5 mL = 0.1 to 0.5 mL
  • 3 mL = 0.5 to 3 mL
  • 12 mL = 3 to 12 mL
  • 30 mL = 12 to 30 mL
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at room temperature
Purification Target Buffer Exchange, Protein
Membrane Cellulose Membrane, Dialysis
Format Cassette
MWCO 10 kDa
Product Line Slide-A-Lyzer
Quantity 6 Cassettes
Sample Volume (Metric) 30 mL
Product Type Dialysis Cassettes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.