missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
SLFN12 Polyclonal antibody specifically detects SLFN12 in Human samples. It is validated for Western Blot, ELISA
Specifications
Specifications
| Antigen | SLFN12 |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | mFluor Violet 610 SE |
| Dilution | Western Blot, ELISA |
| Formulation | 50mM Sodium Borate |
| Gene Alias | schlafen family member 12 |
| Host Species | Rabbit |
| Immunogen | Antibody was raised against a 12 amino acid synthetic peptide from near the carboxy terminus human SLFN12. The immunogen is located within the last 50 amino acids of SLFN12. Amino Acid Squence: FQKNDKKMFKS |
| Purification Method | Peptide affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?