missing translation for 'onlineSavingsMsg'
Learn More

Sampling Systems™ Sleeve Sample Chambers

Product Code. 12947895 Shop All Sampling Systems Products
Click to view available options
Capacity (Metric):
100 mL
150 mL
200 mL
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 12947895

Brand: Sampling Systems™ 1035T200

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Quickly attached to the same handle to allow different sample volumes to be taken. Sample Systems™ Sleeve Sample Chambers are ideal for sampling large volumes at great depths.

  • Made from high quality 316L stainless steel
  • Bright finish
  • Single Point
  • Easy to use

Specifications

Product Type Sample Chamber
Material Stainless Steel (316L)
Capacity (Metric) 200 mL
For Use With (Application) Powder Sampling
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.