missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sleeping Beauty Transposase Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 4 publications
Brand: R&D Systems MAB2798-SP
This item is not returnable.
View return policy
Description
Sleeping Beauty Transposase Monoclonal specifically detects Sleeping Beauty Transposase in Multi-Species samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Sleeping Beauty Transposase | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Detects Sleeping Beauty (SB) Transposase in Western blots. In Immunohistochemistry tests, this antibody stains SB transposase transgenic mouse tissue sections but not wild type mouse tissue sections. | |
| Multi-species | |
| Purified |
| Western Blot, Immunohistochemistry | |
| 324622 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| E. coli-derived recombinant Sleeping Beauty Transposase | |
| 25 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction