Learn More
Invitrogen™ SLCO6A1 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA5116607
Product Code. 17289103
  • £454.00 / 100µL
Description
Positive Samples: SH-SY5Y, 293T, A-549, NCI-H460, Mouse brain Immunogen sequence: VQFAGINEDY DGTGKLGNLT APCNEKCRCS SSIYSSICGR DDIEYFSPCF AGCTYSKAQN QKKMYYNCSC IKEGLITADA EGDFIDARPG KCDAKCYKLP L.
The function remains unknown.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SLCO6A1
Polyclonal
Unconjugated
SLCO6A1
cancer/testis antigen 48; CT48; Gonad-specific transporter; GST; OATP6A1; OATP-I; OATPY; Organic anion-transporting polypeptide 6A1; Organic anion-transporting polypeptide I; SLC21A19; SLCO6A1; Solute carrier family 21 member 19; solute carrier organic anion transporter family member 6A1; solute carrier organic anion transporter family, member 6A1; testicular tissue protein Li 174; testis-specific organic anion transporter
Rabbit
Affinity chromatography
RUO
133482
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Western Blot
2.14 mg/mL
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Q86UG4
SLCO6A1
Recombinant fusion protein containing a sequence corresponding to amino acids 480-580 of human SLCO6A1 (NP_7757593).
100 μL
Primary
Human, Mouse, Rat