missing translation for 'onlineSavingsMsg'
Learn More

SLCO6A1 Antibody, Novus Biologicals™

Product Code. 18682616 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
25 μL
Unit Size:
0.1mL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18682616 25 μL 25µL
18635956 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18682616 Supplier Novus Biologicals Supplier No. NBP24911825ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SLCO6A1 Polyclonal antibody specifically detects SLCO6A1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLCO6A1
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500
Formulation PBS (pH 7.2), 40% Glycerol
Gene Alias Cancer/testis antigen 48, CT48Organic anion-transporting polypeptide I, Gonad-specific transporter, GSTOATP-I, MGC26949, OATP6A1Solute carrier family 21 member 19, OATPY, Organic anion-transporting polypeptide 6A1, SLC21A19, solute carrier organic anion transporter family member 6A1, solute carrier organic anion transporter family, member 6A1, testis-specific organic anion transporter
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to amino acids: AGCTYSKAQNQKKMYYNCSCIKEGLITADAEGDFIDARPGKCDAKCYKLPL
Purification Method Immunogen affinity purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Cancer, Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 133482
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.