missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SLCO5A1 Recombinant Rabbit Monoclonal Antibody (K01_1Y49)

Product Code. 30825789
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30825789 50 μL
30827113 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30825789 Supplier Invitrogen™ Supplier No. MA565146

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

SLCO5A1 Recombinant Monoclonal Antibody for Western Blot

This gene encodes a 12 transmembrane domain protein that is a member of the solute carrier organic anion transporter superfamily. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLCO5A1
Applications Western Blot
Classification Recombinant Monoclonal
Clone K01_1Y49
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 0.05% BSA, 40% glycerol, 150mM NaCl and 0.01% sodium azide; pH 7.4
Gene SLCO5A1
Gene Accession No. Q9H2Y9
Gene Alias 4732454B15; A630033C23Rik; AA407151; LOW QUALITY PROTEIN: solute carrier organic anion transporter family member 5A1; OATP5A1; OATPJ; OATP-J; OATPRP4; OATP-RP4; organic anion transporter polypeptide-related protein 4; SLC21A15; SLCO5A1; solute carrier family 21 (organic anion transporter), member 15; solute carrier family 21 member 15; Solute carrier organic anion transporter family member 5A1; solute carrier organic anion transporter family, member 5A1
Gene Symbols SLCO5A1
Host Species Rabbit
Immunogen Recombinant protein of human SLCO5A1.
Purification Method Protein A/G
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 240726, 81796
Target Species Human, Mouse
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.