missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
SLCO5A1 Polyclonal specifically detects SLCO5A1 in Human samples. It is validated for Western Blot.
Specifications
Specifications
| Antigen | SLCO5A1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS, 2% Sucrose |
| Gene Alias | FLJ39560, member 15, OATP5A1, OATPRP4, organic anion transporter polypeptide-related protein 4, SLC21A15, solute carrier organic anion transporter family, member 5A1 |
| Host Species | Rabbit |
| Immunogen | The immunogen is a synthetic peptide directed towards the middle region of human SLCO5A1 |
| Purification Method | Immunogen affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?