missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
SLCO5A1 Polyclonal Antibody for IHC
Specifications
Specifications
| Antigen | SLCO5A1 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | SLCO5A1 |
| Gene Accession No. | Q9H2Y9 |
| Gene Alias | 4732454B15; A630033C23Rik; AA407151; LOW QUALITY PROTEIN: solute carrier organic anion transporter family member 5A1; OATP5A1; OATPJ; OATP-J; OATPRP4; OATP-RP4; organic anion transporter polypeptide-related protein 4; SLC21A15; SLCO5A1; solute carrier family 21 (organic anion transporter), member 15; solute carrier family 21 member 15; Solute carrier organic anion transporter family member 5A1; solute carrier organic anion transporter family, member 5A1 |
| Gene Symbols | SLCO5A1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
